missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue, TCI America™
Specifications
Specifications
| Melting Point | 222°C |
| Formula Weight | 466.59 |
| Color | Green/Yellow |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Thymol Blue |
Safety and Handling
| RTECSNumber | XP2575000 |
| TSCA | Yes |