missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue Sodium Salt, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS | 62625-21-2 |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Molecular Formula | C27H29NaO5S |
| InChI Key | BAVBEHWEOJMHDS-UHFFFAOYSA-M |
| SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)[O-])C(C)C)C(C)C)O.[Na+] |
| IUPAC Name | sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate |
| Molecular Weight (g/mol) | 488.574 |
| PubChem CID | 23692293 |
| Grade | ACS |
| Show More |
Safety and Handling
| ShelfLife | 60 months |