missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue Indicator, Honeywell Fluka™

Indicator, S. No.: 879

$117.28 - $347.36

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym Thymolsulfonphthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046159
View Documents
Honeywell-Fluka
327285G
5 g Glass Bottle
Each for $117.28
Only null left
 
60046158
View Documents
Honeywell-Fluka
3272825G
25 g Glass Bottle
Each for $347.36
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

76-61-9
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C27H30O5S
MFCD00005869
Thymolsulfonphthalein
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol

Specifications

221°C to 224°C
C27H30O5S
NONH for all modes of transport
Thymolsulfonphthalein
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
466.592
466.59g/mol
1.2 to 2.8 (red to yellow)
5 g
Thymol Blue
76-61-9
MFCD00005869
368036
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
Glass Bottle
0.979g/cm3
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-973-3

SDS
missing translation for 'documents'

missing translation for 'documents'