missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymol Blue, pure, indicator
CAS: 76-61-9 | C27H30O5S | 466.592 g/mol
Supplier: Thermo Scientific Chemicals 151450050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Thymol Blue | |
| 76-61-9 | |
| MFCD00005869 | |
| TB, Thymolsulfonephthalein | |
| PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
| CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
| 466.592 | |
| 466.58 | |
| Glass bottle | |
| Acid range: from pH 1.2 (red) to pH 2.8 (yellow), alkaline range: from pH 8.0 (yellow) to pH 9.6 (blue) | |
| 5 g |
| 221.0°C | |
| C27H30O5S | |
| 15, 9555 | |
| Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions, soluble in alcohol | |
| Authentic | |
| 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol | |
| 65565 | |
| Pure | |
| 700 min. (596nm, in 0.01N NaOH) | |
| Brown to Green | |
| Fine Crystalline Powder |
Chemical Identifiers
| 76-61-9 | |
| 466.592 | |
| PRZSXZWFJHEZBJ-UHFFFAOYSA-N | |
| 65565 | |
| CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
| C27H30O5S | |
| MFCD00005869 | |
| TB, Thymolsulfonephthalein | |
| 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
Safety and Handling
EINECSNumber : 200-973-3
RTECSNumber : XP2575000
TSCA : TSCA
RUO – Research Use Only