missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 2004450
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2004450 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2004450 Supplier Avantor Supplier No. V85602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 76-61-9
Molecular Formula C27H30O5S
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
Molecular Weight (g/mol) 466.592
PubChem CID 65565
Formula Weight 466.6
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 10 g
Chemical Name or Material Thymol Blue
Show More Show Less