missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue, ACS

pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue) | CAS: 76-61-9 | C27H30O5S | 466.592 g/mol

Supplier:  Thermo Scientific Chemicals 01627209

 View more versions of this product

Catalog No. AA1627209


Only null left
Explore available promotions

Description

Description

pH indicator: 1.2 (pink) to 2.8 (yellow) to 9.6 (blue). Thymol Blue is useful as a pH indicator. It is a component of universal indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thymol Blue
76-61-9
MFCD00005869
Thymolsulfonphthalein
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
ACS
Crystalline
223°C (decomposition)
C27H30O5S
14,9400
Soluble in alcohol and dilute alkali solutions; Insoluble in water .
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
466.592
466.6
10 g

Chemical Identifiers

76-61-9
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C27H30O5S
MFCD00005869
Thymolsulfonphthalein
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-973-3

RTECSNumber : XP2575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only