missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue

CAS: 76-61-9 | C27H30O5S | 466.592 g/mol

$82.62 - $268.33

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2137009
View Documents
Thermo Scientific Chemicals
B2137009
10 g
Each for $82.62
Only null left
 
AAB2137018
View Documents
Thermo Scientific Chemicals
B2137018
50 g
Each for $268.33
Only null left
 
Description

Description

A sulfonphthalein dye used as a pH indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfonphthalein dye used as a pH indicator.

Solubility
Solubility in 0.1 N sodium hydroxide solution, almost transparent. Soluble in alcohol, dilute alkali, 0.1% ethanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

76-61-9
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
C27H30O5S
MFCD00005869
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

223°C (decomposition)
C27H30O5S
14,9400
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
466.592
466.6
Thymol Blue
76-61-9
MFCD00005869
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
10 g
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-973-3

RTECSNumber : XP2575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only