missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue, ACS reagent

CAS: 76-61-9 | C27H30O5S | 466.592 g/mol

$79.40 - $131.10

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym TB, Thymolsulfonephthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC420930050
SDS View Documents
Thermo Scientific Chemicals
420930050
5 g Glass bottle
Each for $79.40
Only null left
 
AC420930100
SDS View Documents
Thermo Scientific Chemicals
420930100
10 g Glass bottle
Each for $131.10
Case of 6 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

76-61-9
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C27H30O5S
MFCD00005869
TB, Thymolsulfonephthalein
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol

Specifications

76-61-9
C27H30O5S
15, 9555
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
466.592
466.58
Glass bottle
Brown to Green
Fine Crystalline Powder
Passes Test
MFCD00005869
TB, Thymolsulfonephthalein
Authentic
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
ACS Reagent
Acid range from pH 1.2 (pink) to pH 2.8 (yellow), Alkaline range from pH 8.0 (yellow) to pH 9.2 (blue)
5 g
Thymol Blue, For analysis ACS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only