missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue, 0.08% (w/v) in Methanol, Ricca Chemical

$148.73 - $148.73

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) Mixture
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
8420816
View Documents
Ricca Chemical Company
8420816
500 mL Natural Poly Bottle
Each for $148.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

76-61-9
Mixture
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C27H30O5S
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol

Specifications

76-61-9,67-56-1
100.0,0.1
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
Indicator
Natural Poly Bottle
500 mL
Thymol Blue
98.0,0.1
C27H30O5S
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
Mixture
0.08% W/V
0.08% (w/v) in Methanol
Dark Green
Liquid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
missing translation for 'documents'

missing translation for 'documents'