missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue, 0.08% (w/v) in Methanol, Ricca Chemical
Specifications
Specifications
| CAS | 76-61-9, 67-56-1 |
| CAS Min % | 98.0, 0.1 |
| CAS Max % | 100.0, 0.1 |
| Molecular Formula | C27H30O5S |
| InChI Key | PRZSXZWFJHEZBJ-UHFFFAOYSA-N |
| SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
| IUPAC Name | 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol |
| Molecular Weight (g/mol) | 466.592 |
| PubChem CID | 65565 |
| Concentration or Composition (by Analyte or Components) | 0.08% (w/v) in Methanol |
| Show More |
Safety and Handling
| ShelfLife | 24 months |