missing translation for 'onlineSavingsMsg'
Learn More

Thymidine 98.0+%, TCI America™

Supplier:  TCI America T023325G

Quantity 25 g

 View more versions of this product

Catalog No. T023325G


Only null left

Description

Description

Specifications

Chemical Identifiers

50-89-5
242.23
IQFYYKKMVGJFEH-UHFFFAOYNA-N
5789
1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
C10H14N2O5
MFCD00006537
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
CHEBI:17748
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Thymidine
186°C
C10H14N2O5
MFCD00006537
IQFYYKKMVGJFEH-UHFFFAOYNA-N
1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
5789
242.23
Crystalline Powder
50-89-5
White
25 g
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
242.23
CHEBI:17748
≥98.0% (HPLC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XP2071000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions