missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine, 99+%

Catalog No. AC226741000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226741000 100 g Glass bottle
AC226740050 5 g Glass bottle
AC226740250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226741000 Supplier Thermo Scientific Chemicals Supplier No. 226741000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Absorbance 0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260)
Melting Point 187.0°C to 189.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 159552
Specific Rotation Condition + 18.50 (20.00°C c=3,H2O)
Specific Rotation + 18.50
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, hot ethanol, hot acetone, hot, ethyl acetate and pyridine, sparingly soluble in hot chloroform
Spectrophotometry 98.0 to 102% (lambda max. 267nm,pH 7) molar absorptivity 9600+/-4%,on dry basis
Formula Weight 242.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Thymidine
Show More Show Less
Hazard Category Warning
EINECSNumber 200-070-4

RUO – Research Use Only