missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine, 99+%

CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol

Supplier:  Thermo Scientific Chemicals 226740250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC226740250

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-89-5
242.23
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
CHEBI:17748
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
C10H14N2O5
IQFYYKKMVGJFEH-UHFFFAOYNA-N
5789
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione

Specifications

Thymidine
0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260)
White
99% min. (HPLC)
C10H14N2O5
159552
+ 18.50
Solubility in water: 46g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, hot ethanol, hot acetone, hot, ethyl acetate and pyridine, sparingly soluble in hot chloroform
IQFYYKKMVGJFEH-UHFFFAOYNA-N
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
5789
242.23
Crystalline Powder
50-89-5
187.0°C to 189.0°C
Authentic
Glass bottle
25 g
+ 18.50 (20.00°C c=3,H2O)
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
98.0 to 102% (lambda max. 267nm,pH 7) molar absorptivity 9600+/-4%,on dry basis
CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
242.23
CHEBI:17748
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

EINECSNumber : 200-070-4

SDS
Documents

Documents

Promotions

Promotions