missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine, 99%

Catalog No. p-7052394
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1149306 5 g
AAA1149314 25 g
AAA1149322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1149306 Supplier Thermo Scientific Chemicals Supplier No. A1149306
Only null left

CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol

Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

Solubility
Soluble in water, methanol, hot ethanol, hot acetone, hot ethyl acetate, pyridine, glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
MDL Number MFCD00006537
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 186°C to 190°C
Odor Odorless
Quantity 5 g
Beilstein 89285
Merck Index 14,9397
Solubility Information Soluble in water,methanol,hot ethanol,hot acetone,hot ethyl acetate,pyridine,glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .
Optical Rotation +19° (c=1 in water)
Formula Weight 242.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thymidine
Show More Show Less
EINECSNumber 200-070-4
RTECSNumber XP2071000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only