missing translation for 'onlineSavingsMsg'
Learn More

Thulium powder, -40 mesh, 99.9% (REO)

Catalog No. p-7059198
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4416803 1 g
AA4416806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4416803 Supplier Thermo Scientific Chemicals Supplier No. 04416803
Only null left

CAS: 7440-30-4 | Tm | 168.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 7440-30-4
Molecular Formula Tm
Molecular Weight (g/mol) 168.93
MDL Number MFCD00011281
InChI Key FRNOGLGSGLTDKL-UHFFFAOYSA-N
Synonym tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
PubChem CID 23961
ChEBI CHEBI:33380
IUPAC Name thulium
SMILES [Tm]

Specifications

Color Gray to Silver
Quantity 1 g
Mesh Size −40 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3089
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Thulium powder
Hazard Category H228-H500
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 231-140-2
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only