missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) trifluoromethanesulfonate, 98%

Catalog No. AA4031304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4031304 2 g
AA4031309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4031304 Supplier Thermo Scientific Chemicals Supplier No. 04031304

CAS: 141478-68-4 | C3F9O9S3Tm | 616.124 g/mol

A raw material used in synthesis of chemicals .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141478-68-4
Molecular Formula C3F9O9S3Tm
Molecular Weight (g/mol) 616.124
MDL Number MFCD00209617
InChI Key PBASUZORNBYVFM-UHFFFAOYSA-K
Synonym thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate
PubChem CID 2733936
IUPAC Name thulium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3]

Specifications

Physical Form Powder
Quantity 2 g
Linear Formula Tm(OSO2CF3)3
Solubility Information Soluble in water.
Formula Weight 616.13
Percent Purity 98%
Chemical Name or Material Thulium(III) trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only