missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) bromide, ultra dry, 99.99% (REO)

Catalog No. AA4719603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4719603 1 g
AA4719606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4719603 Supplier Thermo Scientific Chemicals Supplier No. 04719603

CAS: 14456-51-0 | Br3Tm | 408.646 g/mol

Thulium(III) bromide is used for development of advanced ceramic discharge lamps1 and metal halide lamp. Reagent for complexation of lanthanide bromides with aluminum bromide. Reactant for preparation of alkali metal thulium bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14456-51-0
Molecular Formula Br3Tm
Molecular Weight (g/mol) 408.646
MDL Number MFCD00050138
InChI Key HQSWGSFQSCMHFQ-UHFFFAOYSA-K
Synonym thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g
PubChem CID 84454
IUPAC Name thulium(3+);tribromide
SMILES [Br-].[Br-].[Br-].[Tm+3]

Specifications

Boiling Point 1440°C
Melting Point 952°C
Physical Form Beads
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 408.66
Percent Purity 99.99%
Packaging Plastic bottle; Ampouled under argon
Sensitivity Hygroscopic
Chemical Name or Material Thulium(III) bromide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-444-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only