missing translation for 'onlineSavingsMsg'
Learn More

Thulium foil, 0.25mm (0.01in) thick, 99.9% (REO)

Catalog No. p-7048386
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10375FF 25 x 25 mm
AA10375FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10375FF Supplier Thermo Scientific Chemicals Supplier No. 010375FF
Only null left

CAS: 7440-30-4 | Tm | 168.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7440-30-4
Molecular Formula Tm
Molecular Weight (g/mol) 168.93
MDL Number MFCD00011281
InChI Key FRNOGLGSGLTDKL-UHFFFAOYSA-N
Synonym tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
PubChem CID 23961
ChEBI CHEBI:33380
IUPAC Name thulium
SMILES [Tm]

Specifications

Color Silver to Gray
Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.75g/25x25mm
Form Foil
UN Number UN3178
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Thulium foil, 0.25mm (0.01 in.) thick
Hazard Category H228-H260
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-140-2
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only