missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Thrombin, Restriction Grade
Description
- A highly purified preparation qualified to specifically cleave target proteins produced with appropriate vectors
- This preparation is functionally tested for activity with fusion proteins and is free of detectable contaminating proteases
- Thrombin is supplied as a solution in 50% glycerol and includes 10X Thrombin Cleavage Buffer and a Cleavage Control Protein
Specifications
Specifications
| CAS | 9002-04-4 |
| Content And Storage | -20°C |
| Quantity | 50 U |