missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Three Finger Adjustable Clamp, Large

Catalog No. 31501955
Change view
Click to view available options
Size:
Large
For Use With (Equipment):
Accepts objects 0 to 102 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Equipment)
31501955 Large Accepts objects 0 to 102 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31501955 Supplier Synthware™ Supplier No. W181103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Large, two-way adjustable, PVC coated prongs.

Specifications

Type 3-Prong Dual Adjust Clamp
Material Chrome-plated Zinc Alloy
For Use With (Equipment) Accepts objects 0 to 102 mm
For Use With (Application) Organic Chemistry
Length (Metric) 152 mm
Size Large
Size (Metric) Grip 0 to 102 mm
Color Silver
Coating PVC