missing translation for 'onlineSavingsMsg'
Learn More

S-Curve™ Three Compartment Glove Dispenser with Individual Lids, Front Access

Catalog No. 19162424
Change view
Click to view available options
Material:
PETG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19162424 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19162424 Supplier S-Curve™ Supplier No. GD300PETG
Only null left

Three compartment dispenser includes three individually opening lids, with front access holes for easy product dispensing.

  • Three compartment dispenser with individually opening lids
  • Front access holes for easy product dispensing
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 16 x 12 in.
Holds Gloves
Material PETG
Mounting Keyhole Slots
No. of Compartments 3
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.