Learn More
Synthware™ Threaded Top Conical Reaction Vial
GREENER_CHOICE
Heavy walled conical vials designed for high recovery.
Supplier:  Synthware™ V130711
 View more versions of this product
Catalog No. 31502108
Only null left
Description
Description
Specifications
Chemical Identifiers
Clear
Non-sterile
Silicone
Yes
PTFE
1 Ea.
Borosilicate Glass
Single-use
Organic Synthesis
Heavy-wall
Compression Cap
Specifications
Clear
Non-sterile
1 mL
Organic Synthesis
Heavy-wall
Compression Cap
7/10
Borosilicate Glass
Single-use
Silicone
Yes
PTFE
1 Ea.
Product Suggestions
Product Suggestions