missing translation for 'onlineSavingsMsg'
Learn More

Idex Threaded Luer PEEK Adapter

Catalog No. 29008385
Encompass
Change view
Click to view available options
Color:
Black
Natural
For Use With (Equipment):
Female Luer x Female M6 Flat Bottom
Male Luer Lock x Female 10-32 Coned
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
29008385 Natural Male Luer Lock x Female 10-32 Coned
05700336 Black Female Luer x Female M6 Flat Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29008385 Supplier Idex Supplier No. UP P 656

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Convenience of quick connect and disconnect of a luer connection

  • Manufactured from inert polymers

Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.

Specifications

Type Threaded Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 125°C
Material PEEK
Color Natural
For Use With (Equipment) Male Luer Lock x Female 10-32 Coned