missing translation for 'onlineSavingsMsg'
Learn More

Thorin 98.0+%, TCI America™

Catalog No. H02401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H02401G Supplier TCI America Supplier No. H02401G
Only null left

Specifications

CAS 3688-92-4
Molecular Formula C16H11AsN2Na2O10S2
Quantity 1 g
MDL Number MFCD00003888
UN Number 3465
Synonym thoron, thoron ol, 1-2-arsonophenylazo-2-naphthol-3,6-disulfonic acid disodium salt, apans, aspan, disodium 4e-4-2-arsonophenyl hydrazinylidene-3-oxonaphthalene-2,7-disulfonate, thoronol, o-3,6-disulfo-2-hydroxy-1-naphthyl-azo-benzenearsonic acid, 2-2-hydroxy-3,6-disulfo-1-naphthylazo phenylarsonic acid disodium salt, 2-3,6-disulfo-2-hydroxy-1-naphthylazo benzenearsonic acid disodium salt
InChI Key ZQMPNLOZJUWHSA-WTBAQZMLSA-L
SMILES C1=CC=C(C(=C1)NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[As](=O)(O)O.[Na+].[Na+]
IUPAC Name disodium;(4E)-4-[(2-arsonophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 576.289
PubChem CID 6537679
Formula Weight 576.29
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thorin [for Determination of Th, etc.]
Show More Show Less