missing translation for 'onlineSavingsMsg'
Learn More

Thiostrepton, Streptomyces laurentii, 90+%

Catalog No. AAJ6233203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6233203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6233203 Supplier Thermo Scientific Chemicals Supplier No. J6233203

Natural cyclic oligopeptide antibiotic | CAS: 1393-48-2 | C72H85N19O18S5 | 1664.89 g/mol

Thiostrepton exhibits antibiotic activity through blockade of ribosome turnover after an elongation step in protein synthesis by inhibiting both the dissociation of elongation factor G (EF-G) from the ribosome and the release of inorganic phosphate from EF-G following GTP hydrolysis. Thiostrepton is also described to show efficacy against cancer cell growth through downregulation of the forkhead box M1 (FOXM1) transcription factor, commonly overexpressed in many malignancies. Thiostrepton is an inhibitor of FOXM1. Thiostrepton has been used as a selectable marker for recombinant Streptomyces cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiostrepton exhibits antibiotic activity through blockade of ribosome turnover after an elongation step in protein synthesis by inhibiting both the dissociation of elongation factor G (EF-G) from the ribosome and the release of inorganic phosphate from EF-G following GTP hydrolysis. Thiostrepton is also described to show efficacy against cancer cell growth through downregulation of the forkhead box M1 (FOXM1) transcription factor, commonly overexpressed in many malignancies. Thiostrepton is an inhibitor of FOXM1. Thiostrepton has been used as a selectable marker for recombinant Streptomyces cultures.

Solubility
Soluble in DMSO or chloroform

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.

Specifications

Type Thiostrepton
Physical Form Powder
CAS 1393-48-2
Color White to Yellow
Quantity 1 g
Molecular Formula C72H85N19O18S5
MDL Number MFCD00135828,MFCD00135828
Merck Index 14,9364
Synonym Bryamycin; Thiactin
Solubility Information Soluble in DMSO or chloroform
InChI Key NSFFHOGKXHRQEW-DVRIZHICNA-N
SMILES CCC(C)C1NC2C=CC3=C(C=C(N=C3C2O)C(=O)OC(C)C2NC(=O)C3=CSC(=N3)C(NC(=O)C3CSC(=N3)\C(NC(=O)C(NC(=O)C3=CSC(=N3)C3(CCC(=NC3C3=CSC2=N3)C2=NC(=CS2)C(=O)NC(=C)C(=O)NC(=C)C(N)=O)NC(=O)C(C)NC(=O)C(=C)NC(=O)C(C)NC1=O)C(C)O)=C/C)C(C)(O)C(C)O)C(C)O
IUPAC Name 2-({2-[(11E)-37-(butan-2-yl)-18-(2,3-dihydroxybutan-2-yl)-11-ethylidene-59-hydroxy-8,60-bis(1-hydroxyethyl)-26,40,46-trimethyl-43-methylidene-6,9,16,23,28,38,41,44,47-nonaoxo-27-oxa-3,13,20,56-tetrathia-7,10,17,24,30,36,39,42,45,48,52,58,62,63,64-pentadecaazanonacyclo[23.23.9.2²⁹,³².1²,⁵.1¹²,¹⁵.1¹⁹,²².1³¹,³⁵.1⁵⁴,⁵⁷.0¹,⁵³]tetrahexaconta-2(64),4,12(63),19(62),21,29(61),30,32(60),33,51,54,57-dodecaen-51-yl]-1,3-thiazol-4-yl}formamido)-N-(1-carbamoyleth-1-en-1-yl)prop-2-enamide
Molecular Weight (g/mol) 1664.89
PubChem CID 122173112
Formula Weight 1664.89
Percent Purity ≥90%
Chemical Name or Material Thiostrepton, Streptomyces laurentii
Show More Show Less
EINECSNumber 215-734-9
RTECSNumber XN6300100
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only