missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Thiostrepton, Calbiochem™,

Supplier:  MilliporeSigma™ 59822610GM

 View more versions of this product

Catalog No. 59822610GM


Only null left
Explore available promotions

Description

Description

Recommended Storage

2°C to 8°C, Following reconstitution, aliquot and freeze (−20°C). Stock solutions are Stable for up to 6 months at −20°C

Specifications

Specifications

1393-48-2
Glass Bottle
MFCD00135828,MFCD00135828
thiostrepton
CCC(C)C1NC2C=CC3=C(C=C(N=C3C2O)C(=O)OC(C)C2NC(=O)C3=CSC(=N3)C(NC(=O)C3CSC(=N3)\C(NC(=O)C(NC(=O)C3=CSC(=N3)C3(CCC(=NC3C3=CSC2=N3)C2=NC(=CS2)C(=O)NC(=C)C(=O)NC(=C)C(N)=O)NC(=O)C(C)NC(=O)C(=C)NC(=O)C(C)NC1=O)C(C)O)=C/C)C(C)(O)C(C)O)C(C)O
1664.89
Powder
White
C72H85N19O18S5
10 g
NSFFHOGKXHRQEW-DVRIZHICNA-N
2-({2-[(11E)-37-(butan-2-yl)-18-(2,3-dihydroxybutan-2-yl)-11-ethylidene-59-hydroxy-8,60-bis(1-hydroxyethyl)-26,40,46-trimethyl-43-methylidene-6,9,16,23,28,38,41,44,47-nonaoxo-27-oxa-3,13,20,56-tetrathia-7,10,17,24,30,36,39,42,45,48,52,58,62,63,64-pentadecaazanonacyclo[23.23.9.2²⁹,³².1²,⁵.1¹²,¹⁵.1¹⁹,²².1³¹,³⁵.1⁵⁴,⁵⁷.0¹,⁵³]tetrahexaconta-2(64),4,12(63),19(62),21,29(61),30,32(60),33,51,54,57-dodecaen-51-yl]-1,3-thiazol-4-yl}formamido)-N-(1-carbamoyleth-1-en-1-yl)prop-2-enamide
122173112

Chemical Identifiers

1393-48-2
1664.89
NSFFHOGKXHRQEW-DVRIZHICNA-N
122173112
CCC(C)C1NC2C=CC3=C(C=C(N=C3C2O)C(=O)OC(C)C2NC(=O)C3=CSC(=N3)C(NC(=O)C3CSC(=N3)\C(NC(=O)C(NC(=O)C3=CSC(=N3)C3(CCC(=NC3C3=CSC2=N3)C2=NC(=CS2)C(=O)NC(=C)C(=O)NC(=C)C(N)=O)NC(=O)C(C)NC(=O)C(=C)NC(=O)C(C)NC1=O)C(C)O)=C/C)C(C)(O)C(C)O)C(C)O
C72H85N19O18S5
MFCD00135828,MFCD00135828
thiostrepton
2-({2-[(11E)-37-(butan-2-yl)-18-(2,3-dihydroxybutan-2-yl)-11-ethylidene-59-hydroxy-8,60-bis(1-hydroxyethyl)-26,40,46-trimethyl-43-methylidene-6,9,16,23,28,38,41,44,47-nonaoxo-27-oxa-3,13,20,56-tetrathia-7,10,17,24,30,36,39,42,45,48,52,58,62,63,64-pentadecaazanonacyclo[23.23.9.2²⁹,³².1²,⁵.1¹²,¹⁵.1¹⁹,²².1³¹,³⁵.1⁵⁴,⁵⁷.0¹,⁵³]tetrahexaconta-2(64),4,12(63),19(62),21,29(61),30,32(60),33,51,54,57-dodecaen-51-yl]-1,3-thiazol-4-yl}formamido)-N-(1-carbamoyleth-1-en-1-yl)prop-2-enamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C, Following reconstitution, aliquot and freeze (−20°C). Stock solutions are Stable for up to 6 months at −20°C

SDS
Documents

Documents

Promotions

Promotions