missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide, 98+%

Catalog No. p-200281
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138901000 100 g Plastic bottle
AC138905000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138901000 Supplier Thermo Scientific Chemicals Supplier No. 138901000

CAS: 79-19-6 | CH5N3S | 91.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-19-6
Molecular Formula CH5N3S
Molecular Weight (g/mol) 91.13
MDL Number MFCD00007620
InChI Key BRWIZMBXBAOCCF-UHFFFAOYSA-N
Synonym thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
PubChem CID 2723789
IUPAC Name aminothiourea
SMILES C(=S)(N)NN

Specifications

Melting Point 175.0°C to 180.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula H2NCSNHNH2
Quantity 100 g
Beilstein 03,195
Fieser 01,1164
Merck Index 15,9514
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 91.13
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Thiosemicarbazide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
EINECSNumber 201-184-7
RTECSNumber VT4200000
TSCA TSCA

RUO – Research Use Only