Learn More
Thiosemicarbazide, 98%
CAS: 79-19-6 | CH5N3S | 91.132 g/mol
Supplier: Thermo Scientific Chemicals A146300E
Description
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.
Solubility
Soluble in water, ethanol, alcohols and hot methanol.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
| Thiosemicarbazide | |
| ∼180°C (decomposition) | |
| CH5N3S | |
| 2500 g | |
| 506320 | |
| thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
| BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
| aminothiourea | |
| 2723789 | |
| 99% |
| 79-19-6 | |
| Odorless | |
| MFCD00007620 | |
| UN2811 | |
| 14,9361 | |
| Soluble in water,ethanol,alcohols and hot methanol. | |
| C(=S)(N)NN | |
| 91.132 | |
| 91.14 |
Chemical Identifiers
| 79-19-6 | |
| 91.132 | |
| BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
| 2723789 | |
| C(=S)(N)NN |
| CH5N3S | |
| MFCD00007620 | |
| thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
| aminothiourea |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P264b-P270-P301+P310-P330-P501c
H300
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 201-184-7
RTECSNumber : VT4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only