missing translation for 'onlineSavingsMsg'
Learn More

Thiophosphoryl Chloride 98.0+%, TCI America™

Catalog No. T189525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T189525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T189525G Supplier TCI America Supplier No. T189525G
Only null left

Chemical Identifiers

CAS 3982-91-0
Molecular Formula Cl3PS
Molecular Weight (g/mol) 169.384
MDL Number MFCD00011504
InChI Key WQYSXVGEZYESBR-UHFFFAOYSA-N
Synonym Phosphorus Thiochloride
PubChem CID 19883
IUPAC Name trichloro(sulfanylidene)-$l^{5}-phosphane
SMILES P(=S)(Cl)(Cl)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 1837
Formula Weight 169.38
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Thiophosphoryl Chloride
EINECSNumber (1)-0251
RTECSNumber XN2930000
TSCA Yes