missing translation for 'onlineSavingsMsg'
Learn More

Thiophosgene, 85%

Catalog No. p-3737588
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151640250 25 g Glass bottle
AC151641000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151640250 Supplier Thermo Scientific Chemicals Supplier No. 151640250
Only null left

CAS: 463-71-8 | CCl2S | 114.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 463-71-8
MDL Number MFCD00004918
InChI Key ZWZVWGITAAIFPS-UHFFFAOYSA-N
Synonym thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
PubChem CID 10040
ChEBI CHEBI:29366
IUPAC Name thiocarbonyl dichloride
SMILES C(=S)(Cl)Cl

Specifications

Boiling Point 73.0°C
Color Orange-Red to Red
Physical Form Liquid
Quantity 25 g
Assay Percent Range 80% min. (GC)
Molecular Formula CCl2S
Linear Formula ClCSCl
Beilstein 03,134
Fieser 05,667; 09,466
Flash Point 62°C
Solubility Information Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
Molecular Weight (g/mol) 114.98
Formula Weight 114.98
Percent Purity 85%
Packaging Glass bottle
Refractive Index 1.5442
Specific Gravity 1.508
Density 1.5080g/mL
Chemical Name or Material Thiophosgene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
If skin i
EINECSNumber 207-341-6

RUO – Research Use Only