missing translation for 'onlineSavingsMsg'
Learn More

Thiophene 98.0+%, TCI America™

Catalog No. T022325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022325ML 25 mL
T0223500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T022325ML Supplier TCI America Supplier No. T022325ML

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1

Specifications

Melting Point -38°C
Color Colorless
Boiling Point 84°C
Quantity 25 mL
UN Number 2414
Formula Weight 84.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiophene
EINECSNumber (9)-0810
RTECSNumber XM7350000
TSCA Yes