missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene, 99%
Description
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.
Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.
Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | -38°C |
| Density | 1.066 |
| Boiling Point | 83°C to 85°C |
| Flash Point | -9°C (16°F) |
| Odor | Stench |
| Refractive Index | 1.528 |
| Quantity | 500 g |
| UN Number | UN2414 |
| Beilstein | 103222 |
| Merck Index | 14,9353 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H319 |
| Hazard Statement | GHS H StatementH225-H331-H302-H312-H315-H319Highly flammable liquid and vapour.Toxic if inhaled.Harmful if swallowed.Harmful in contact with skin.Causes skin irritation.Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: THIOPHENE |
| EINECSNumber | 203-729-4 |
| RTECSNumber | XM7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only