missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99%

Catalog No. AAA1394118
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1394118 50 g
AAA1394130 250 g
AAA1394136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1394118 Supplier Thermo Scientific Chemicals Supplier No. A1394118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-02-1 | C4H4S | 84.14 g/mol

Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1

Specifications

Melting Point -38°C
Density 1.066
Boiling Point 83°C to 85°C
Flash Point -9°C (16°F)
Odor Stench
Refractive Index 1.528
Quantity 50 g
UN Number UN2414
Beilstein 103222
Merck Index 14,9353
Solubility Information Miscible with ethyl alcohol,diethyl ether and other organic solvents. Immiscible with water.
Formula Weight 84.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiophene
Show More Show Less
Hazard Category H225-H302-H319
Hazard Statement GHS H Statement

H225-H331-H302-H312-H315-H319

Highly flammable liquid and vapour.

Toxic if inhaled.

Harmful if swallowed.

Harmful in contact with skin.

Causes skin irritation.

Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: THIOPHENE
EINECSNumber 203-729-4
RTECSNumber XM7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only