missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99+%, extra pure

Catalog No. p-200206
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138821000 100 g Glass bottle
AC138825000 500 g Glass bottle
AC138820025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138821000 Supplier Thermo Scientific Chemicals Supplier No. 138821000
Only null left

CAS: 110-02-1 | C4H4S | 84.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1

Specifications

Melting Point -38.0°C
Density 1.0650g/mL
Boiling Point 84.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290
Quantity 100 g
Beilstein 17, 29
Specific Gravity 1.065
Merck Index 15, 9506
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 84.14
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Thiophene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 203-729-4
RTECSNumber XM7350000
TSCA TSCA

RUO – Research Use Only