Learn More
Thiophene-3-carboxaldehyde, 96%
CAS: 498-62-4 | C5H4OS | 112.146 g/mol
Supplier: Thermo Scientific Chemicals A1462818
Description
Thiophene-3-carboxaldehyde has been used in the synthesis of series of 4-substituted 2-thiophenesulfonamides, acetal and ketal derivatives of 4?-demethylepipodophyllotoxin-β-D-glucoside and epipodophyllotoxin-β-D-glucoside. Also has been used to produce 1,2-di-3-thienyl-2-hydroxyethanone (3,3?-thenoin), 3-thienyl symmetric analog of benzoin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiophene-3-carboxaldehyde has been used in the synthesis of series of 4-substituted 2-thiophenesulfonamides, acetal and ketal derivatives of 4′-demethylepipodophyllotoxin-β-D-glucoside and epipodophyllotoxin-β-D-glucoside. Also has been used to produce 1,2-di-3-thienyl-2-hydroxyethanone (3,3′-thenoin), 3-thienyl symmetric analog of benzoin.
Solubility
Not miscible in water.
Notes
It is sensitive to air. Incompatible with oxidizing agents and heat.
Specifications
| Thiophene-3-carboxaldehyde | |
| 1.28 | |
| 64°C (147°F) | |
| 1.584 | |
| 50 g | |
| Air sensitive | |
| Not miscible in water. | |
| C1=CSC=C1C=O | |
| 112.146 | |
| CHEBI:87611 | |
| 96% |
| 498-62-4 | |
| 194°C to 196°C | |
| C5H4OS | |
| MFCD00005466 | |
| 105889 | |
| 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde | |
| RBIGKSZIQCTIJF-UHFFFAOYSA-N | |
| thiophene-3-carbaldehyde | |
| 68135 | |
| 112.15 |
Chemical Identifiers
| 498-62-4 | |
| 112.146 | |
| RBIGKSZIQCTIJF-UHFFFAOYSA-N | |
| 68135 | |
| thiophene-3-carbaldehyde |
| C5H4OS | |
| MFCD00005466 | |
| 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde | |
| CHEBI:87611 | |
| C1=CSC=C1C=O |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 207-865-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only