missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carbonitrile, 98%

Catalog No. AAL1311506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1311506 5 g
AAL1311514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1311506 Supplier Thermo Scientific Chemicals Supplier No. L1311506

CAS: 1641-09-4 | C5H3NS | 109.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1641-09-4
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00151852
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
PubChem CID 74231
IUPAC Name thiophene-3-carbonitrile
SMILES C1=CSC=C1C#N

Specifications

Density 1.2
Boiling Point 205°C
Flash Point 86°C (186°F)
Odor Recognizable
Refractive Index 1.5615
Quantity 5 g
UN Number UN3276
Beilstein 107104
Formula Weight 109.15
Percent Purity 98%
Chemical Name or Material Thiophene-3-carbonitrile
Show More Show Less
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 216-687-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only