missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-ethylamine, 98%, Thermo Scientific™

$69.29

Chemical Identifiers

CAS 30433-91-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.21
MDL Number MFCD00051495
InChI Key HVLUYXIJZLDNIS-UHFFFAOYSA-N
Synonym thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
PubChem CID 116521
IUPAC Name 2-thiophen-2-ylethanamine
SMILES C1=CSC(=C1)CCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC354270010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

30433-91-1
127.21
HVLUYXIJZLDNIS-UHFFFAOYSA-N
116521
C1=CSC(=C1)CCN
C6H9NS
MFCD00051495
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
2-thiophen-2-ylethanamine

Specifications

30433-91-1
Authentic
C6H9NS
MFCD00051495
HVLUYXIJZLDNIS-UHFFFAOYSA-N
2-thiophen-2-ylethanamine
116521
98%
103.0°C to 105.0°C (15.0 mmHg)
97.5% min. (GC)
1.5490 to 1.5510
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
C1=CSC(=C1)CCN
127.21
127.21
Thiophene-2-ethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 250-196-9

SDS
Documents

Documents

RUO – Research Use Only