Learn More
Thiophene-2-carboxylic acid, 99%
CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
Supplier: Thermo Scientific Chemicals A1251436
Description
A hypoglycemic, anti-lipolytic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Thiophene-2-carboxylic acid | |
| 126°C to 130°C | |
| 259°C to 261°C | |
| MFCD00005437 | |
| 110150 | |
| 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
| OC(=O)C1=CC=CS1 | |
| 128.15 | |
| CHEBI:71241 | |
| 99% |
| 527-72-0 | |
| 1.42 | |
| C5H4O2S | |
| 500 g | |
| 14,9354 | |
| QERYCTSHXKAMIS-UHFFFAOYSA-N | |
| thiophene-2-carboxylic acid | |
| 10700 | |
| 128.15 |
Chemical Identifiers
| 527-72-0 | |
| 128.15 | |
| QERYCTSHXKAMIS-UHFFFAOYSA-N | |
| 10700 | |
| thiophene-2-carboxylic acid |
| C5H4O2S | |
| MFCD00005437 | |
| 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
| CHEBI:71241 | |
| OC(=O)C1=CC=CS1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
EINECSNumber : 208-423-4
RTECSNumber : XM8330200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only