missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxylic acid, 99%

Catalog No. AAA1251414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1251414 25 g
AAA1251422 100 g
AAA1251436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1251414 Supplier Thermo Scientific Chemicals Supplier No. A1251414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol

A hypoglycemic, anti-lipolytic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 527-72-0
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005437
InChI Key QERYCTSHXKAMIS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid
PubChem CID 10700
ChEBI CHEBI:71241
IUPAC Name thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=CS1

Specifications

Melting Point 126°C to 130°C
Density 1.42
Boiling Point 259°C to 261°C
Quantity 25 g
Beilstein 110150
Merck Index 14,9354
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material Thiophene-2-carboxylic acid
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-423-4
RTECSNumber XM8330200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only