missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxamide, 99%

Catalog No. p-7022436
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1521906 5 g
AAA1521914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1521906 Supplier Thermo Scientific Chemicals Supplier No. A1521906
Only null left

CAS: 5813-89-8 | C5H5NOS | 127.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5813-89-8
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00010425
InChI Key DENPQNAWGQXKCU-UHFFFAOYSA-N
Synonym 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
PubChem CID 22063
IUPAC Name thiophene-2-carboxamide
SMILES C1=CSC(=C1)C(=O)N

Specifications

Melting Point 178°C to 183°C
Quantity 5 g
Beilstein 110153
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material Thiophene-2-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber XM8290000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only