Learn More
Thiophene-2-carboxaldoxime, 98+%
CAS: 29683-84-9 | C5H5NOS | 127.16 g/mol
Supplier: Thermo Scientific Chemicals L1096114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Thiophene-2-carboxaldoxime | |
| 132°C to 136°C | |
| MFCD00022509 | |
| 108676 | |
| GASLBPLHYRZLLT-GQCTYLIASA-N | |
| (NE)-N-(thiophen-2-ylmethylidene)hydroxylamine | |
| 6876527 | |
| ≥98% |
| 29683-84-9 | |
| C5H5NOS | |
| 25 g | |
| thiophene-2-carbaldehyde oxime, 2-thiophenecarboxaldehyde, oxime, thiophene-2-carboxaldoxime, thiophene-2-aldoxime, 2-thiophenealdehyde oxime, e-n-thiophen-2-ylmethylidene hydroxylamine, e-thiophene-2-carbaldehyde oxime, thiophen-2-aldoxim, pubchem14034, 2-formylthiophene oxime | |
| O\N=C\C1=CC=CS1 | |
| 127.16 | |
| 127.17 |
Chemical Identifiers
| 29683-84-9 | |
| 127.16 | |
| GASLBPLHYRZLLT-GQCTYLIASA-N | |
| 6876527 | |
| O\N=C\C1=CC=CS1 |
| C5H5NOS | |
| MFCD00022509 | |
| thiophene-2-carbaldehyde oxime, 2-thiophenecarboxaldehyde, oxime, thiophene-2-carboxaldoxime, thiophene-2-aldoxime, 2-thiophenealdehyde oxime, e-n-thiophen-2-ylmethylidene hydroxylamine, e-thiophene-2-carbaldehyde oxime, thiophen-2-aldoxim, pubchem14034, 2-formylthiophene oxime | |
| (NE)-N-(thiophen-2-ylmethylidene)hydroxylamine |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 249-778-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only