missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxaldoxime, 98+%

Catalog No. AAL1096106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1096106 5 g
AAL1096114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1096106 Supplier Thermo Scientific Chemicals Supplier No. L1096106
Only null left

CAS: 29683-84-9 | C5H5NOS | 127.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29683-84-9
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.16
MDL Number MFCD00022509
InChI Key GASLBPLHYRZLLT-GQCTYLIASA-N
Synonym thiophene-2-carbaldehyde oxime, 2-thiophenecarboxaldehyde, oxime, thiophene-2-carboxaldoxime, thiophene-2-aldoxime, 2-thiophenealdehyde oxime, e-n-thiophen-2-ylmethylidene hydroxylamine, e-thiophene-2-carbaldehyde oxime, thiophen-2-aldoxim, pubchem14034, 2-formylthiophene oxime
PubChem CID 6876527
IUPAC Name (NE)-N-(thiophen-2-ylmethylidene)hydroxylamine
SMILES O\N=C\C1=CC=CS1

Specifications

Melting Point 132°C to 136°C
Quantity 5 g
Beilstein 108676
Formula Weight 127.17
Percent Purity ≥98%
Chemical Name or Material Thiophene-2-carboxaldoxime
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 249-778-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only