missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-boronic acid pinacol ester, 98%

CAS: 193978-23-3 | C10H15BO2S | 210.098 g/mol

Supplier:  Thermo Scientific Chemicals H6381906

 View more versions of this product

Catalog No. AAH6381906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thiophene-2-boronic acid pinacol ester
65°C to 70°C
MFCD05663878
2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2
210.098
228.12
Solid
193978-23-3
C10H15BO2S
5 g
FFZHICFAHSDFKZ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
10703628
98%

Chemical Identifiers

193978-23-3
210.098
FFZHICFAHSDFKZ-UHFFFAOYSA-N
10703628
B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2
C10H15BO2S
MFCD05663878
2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only