Learn More
Thiophene-2-boronic acid, 97%
Description
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 128°C to 131°C |
| Quantity | 1 g |
| Beilstein | 112375 |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 127.96 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Thiophene-2-boronic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H StatementH315-H319-H335Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only