missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene-2-acetyl Chloride 98.0+%, TCI America™
Supplier: TCI America T1751250G
Specifications
| Thiophene-2-acetyl Chloride | |
| Brown-Yellow | |
| C6H5ClOS | |
| 250 g | |
| 2-thiopheneacetyl chloride, 2-thiophen-2-yl acetyl chloride, 2-thienylacetyl chloride, 2-thiophene acetyl chloride, thiophene-2-acetyl chloride, 2-2-thienyl acetyl chloride, thiopheneacetyl chloride, 2-thiopheneacetylchloride, thienylacetyl chloride, thien-2-ylacetyl chloride | |
| C1=CSC(=C1)CC(=O)Cl | |
| 160.615 | |
| 160.62 | |
| Liquid |
| 39098-97-0 | |
| 106°C | |
| MFCD00005456 | |
| 3265 | |
| AJYXPNIENRLELY-UHFFFAOYSA-N | |
| 2-thiophen-2-ylacetyl chloride | |
| 162362 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 39098-97-0 | |
| 160.615 | |
| AJYXPNIENRLELY-UHFFFAOYSA-N | |
| 162362 | |
| C1=CSC(=C1)CC(=O)Cl |
| C6H5ClOS | |
| MFCD00005456 | |
| 2-thiopheneacetyl chloride, 2-thiophen-2-yl acetyl chloride, 2-thienylacetyl chloride, 2-thiophene acetyl chloride, thiophene-2-acetyl chloride, 2-2-thienyl acetyl chloride, thiopheneacetyl chloride, 2-thiopheneacetylchloride, thienylacetyl chloride, thien-2-ylacetyl chloride | |
| 2-thiophen-2-ylacetyl chloride |
Safety and Handling
EINECSNumber : (5)-5326
TSCA : Yes