missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-acetyl Chloride 98.0+%, TCI America™

Catalog No. T175125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T175125G 25 g
T1751250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T175125G Supplier TCI America Supplier No. T175125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39098-97-0
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.615
MDL Number MFCD00005456
InChI Key AJYXPNIENRLELY-UHFFFAOYSA-N
Synonym 2-thiopheneacetyl chloride, 2-thiophen-2-yl acetyl chloride, 2-thienylacetyl chloride, 2-thiophene acetyl chloride, thiophene-2-acetyl chloride, 2-2-thienyl acetyl chloride, thiopheneacetyl chloride, 2-thiopheneacetylchloride, thienylacetyl chloride, thien-2-ylacetyl chloride
PubChem CID 162362
IUPAC Name 2-thiophen-2-ylacetyl chloride
SMILES C1=CSC(=C1)CC(=O)Cl

Specifications

Color Brown-Yellow
Boiling Point 106°C
Quantity 25 g
UN Number 3265
Formula Weight 160.62
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Thiophene-2-acetyl Chloride
EINECSNumber (5)-5326
TSCA Yes