missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-acetonitrile 98.0+%, TCI America™

Catalog No. p-7136436
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07995ML 5 mL
T079925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07995ML Supplier TCI America Supplier No. T07995ML

Chemical Identifiers

CAS 20893-30-5
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.173
MDL Number MFCD00005453
InChI Key CLSHQIDDCJTHAJ-UHFFFAOYSA-N
Synonym 2-thiopheneacetonitrile, 2-thiophen-2-yl acetonitrile, 2-thienylacetonitrile, thiophene-2-acetonitrile, 2-cyanomethyl thiophene, 2-cyanomethylthiophene, 2-thiophene-acetonitrile, thiophen-2-yl-acetonitrile, thien-2-ylacetonitrile, thiophen-2-ylacetonitrile
PubChem CID 72880
ChEBI CHEBI:27382
IUPAC Name 2-thiophen-2-ylacetonitrile
SMILES C1=CSC(=C1)CC#N

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 mL
UN Number 3276
Formula Weight 123.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiophene-2-acetonitrile