missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2,5-diboronic acid bis(pinacol) ester, 97%

Catalog No. p-7034077
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5172503 1 g
AAH5172506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5172503 Supplier Thermo Scientific Chemicals Supplier No. H5172503

CAS: 175361-81-6 | C16H26B2O4S | 336.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175361-81-6
Molecular Formula C16H26B2O4S
Molecular Weight (g/mol) 336.06
MDL Number MFCD09800563
InChI Key AOJXAKMKFDBHHT-UHFFFAOYSA-N
Synonym 2,5-bis-thiopheneboronic acid pinacol ester, 2,5-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2,5-diboronic acid bis pinacol ester, thiophene-2,5-diboronicacidbis pinacol ester, bis 4,4,5,5-tetramethyl-1,3-dioxaborolane-2,5-thiophene, 2,5-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl thiophene, 2,2'-thiene-2,5-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl-1,3,2-dioxaborolane
PubChem CID 23058061
IUPAC Name 4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 216°C to 220°C
Quantity 1 g
Formula Weight 336.06
Percent Purity 97%
Chemical Name or Material Thiophene-2,5-diboronic acid bis(pinacol) ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only