missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. T21935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21935G 5 g
T219325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21935G Supplier TCI America Supplier No. T21935G

Chemical Identifiers

CAS 39093-93-1
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.181
InChI Key NDOVLWQBFFJETK-UHFFFAOYSA-N
Synonym 1,1-Dioxothiomorpholine
PubChem CID 6484228
IUPAC Name 1,4-thiazinane 1,1-dioxide
SMILES C1CS(=O)(=O)CCN1

Specifications

Melting Point 70°C
Color White
Boiling Point 155°C
Quantity 5 g
Formula Weight 135.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiomorpholine 1,1-Dioxide