missing translation for 'onlineSavingsMsg'
Learn More

Thiomichler's Ketone 97.0+%, TCI America™

Catalog No. A52011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52011G Supplier TCI America Supplier No. A52011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Sensitive spectrophotometric reagent for Au, Ag, Hg and Pd, use for the determination of residual chlorine]

Chemical Identifiers

CAS 1226-46-6
Molecular Formula C17H20N2S
Molecular Weight (g/mol) 284.421
MDL Number MFCD00040477
InChI Key KFUJUTFTRXYQMG-UHFFFAOYSA-N
Synonym 4,4′C-Bis(dimethylamino)thiobenzophenone, N,N,N′C,N′C-Tetramethyl-4,4′C-diaminothiobenzophenone
PubChem CID 71045
IUPAC Name bis[4-(dimethylamino)phenyl]methanethione
SMILES CN(C)C1=CC=C(C=C1)C(=S)C2=CC=C(C=C2)N(C)C

Specifications

Melting Point 204°C
Color Purple-Red
Quantity 1 g
Formula Weight 284.42
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiomichler′s Ketone [Sensitive spectrophotometric reagent for Au, Ag, Hg and Pd, use for the